Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ketoisophorone | 1125-21-9 | MFCD00043119 | 99.9% | 152.19 g·mol⁻¹ | C9H12O2 | 100 G
SDP

Catalog No. 5000242353
Encompass_Preferred

Ketoisophorone (4-Oxoisophorone) is a small-molecule synthesis intermediate used as a building block in organic synthesis, flavor and fragrance chemistry, and carotenoid synthesis. Supplied for research use only, it is provided at high purity for laboratory applications.

  • High purity: 99.9%
  • Molecular formula C9H12O2; molecular weight 152.19 g·mol⁻¹
  • CAS number 1125-21-9
  • Commonly used as a synthesis intermediate and building block
  • Intended for research use only
  • Available in multiple pack sizes including 100 g

Catalog No. 50-002-42353 Supplier Medchemexpress LLC Supplier No. HY107832100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ketoisophorone (4-Oxoisophorone) is a small-molecule synthesis intermediate used as a building block in organic synthesis, flavor and fragrance chemistry, and carotenoid synthesis. Supplied for research use only, it is provided at high purity for laboratory applications.

  • High purity: 99.9%
  • Molecular formula C9H12O2; molecular weight 152.19 g·mol⁻¹
  • CAS number 1125-21-9
  • Commonly used as a synthesis intermediate and building block
  • Intended for research use only
  • Available in multiple pack sizes including 100 g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.