Learn More
Medchemexpress LLC Ketoisophorone (4-oxoisophorone) | 1125-21-9 | 99.9% | 152.19 g/mol | C9H12O2 | 25 G
SDP
Catalog No. 5000244491
Encompass_Preferred
Ketoisophorone (4-oxoisophorone) is a high-purity small-molecule synthesis intermediate (CAS 1125-21-9) used as a building block in organic synthesis, including the preparation of carotenoids and flavor compounds. It is supplied as a stable material suitable for laboratory research and specialty chemical manufacturing.
  • High purity suitable for synthesis and research (99.9%).
  • Well-defined chemical formula C9H12O2 and molecular weight 152.19 g/mol.
  • Useful intermediate for carotenoid and flavor compound synthesis.
  • Available in a convenient 25 G pack for laboratory use.
  • Certificate of Analysis and safety data sheet available for quality and handling.
Catalog No. 50-002-44491 Supplier Medchemexpress LLC Supplier No. HY10783225G
Ketoisophorone (4-oxoisophorone) is a high-purity small-molecule synthesis intermediate (CAS 1125-21-9) used as a building block in organic synthesis, including the preparation of carotenoids and flavor compounds. It is supplied as a stable material suitable for laboratory research and specialty chemical manufacturing.
  • High purity suitable for synthesis and research (99.9%).
  • Well-defined chemical formula C9H12O2 and molecular weight 152.19 g/mol.
  • Useful intermediate for carotenoid and flavor compound synthesis.
  • Available in a convenient 25 G pack for laboratory use.
  • Certificate of Analysis and safety data sheet available for quality and handling.