Learn More
Apexbio Technology LLC Ketoisovaleric acid 759-05-7 500mg
SDP
Catalog No. 5000091191
Encompass_Preferred
Ketoisovaleric acid (CAS 759-05-7) is a branched-chain keto acid produced during the catabolism of valine It accumulates in conditions such as maple syrup urine disease (MSUD) a metabolic disorder resulting from deficiency of the branched-chain -keto acid dehydrogenase complex Elevated levels of ketoisovaleric acid have been implicated in promoting oxidative stress and metabolic acidosis due to impaired amino acid breakdown This metabolite is utilized in research investigating metabolic dysregulation oxidative damage and the pathophysiology of organic acidemias and neurological dysfunction in inherited metabolic diseases
Catalog No. 50-000-91191 Supplier Apexbio Technology LLC Supplier No. B7892500
Add to Cart
Add to Cart
Ketoisovaleric acid (CAS 759-05-7) is a branched-chain keto acid produced during the catabolism of valine It accumulates in conditions such as maple syrup urine disease (MSUD) a metabolic disorder resulting from deficiency of the branched-chain -keto acid dehydrogenase complex Elevated levels of ketoisovaleric acid have been implicated in promoting oxidative stress and metabolic acidosis due to impaired amino acid breakdown This metabolite is utilized in research investigating metabolic dysregulation oxidative damage and the pathophysiology of organic acidemias and neurological dysfunction in inherited metabolic diseases