Learn More
Nova Biomedical Corporation Nova Max Ketone/Glucose Control Solution – #49640
Supplier:  Nova Biomedical Corporation 49640
Encompass
The Quality Control Solution ensures accurate operation of the Nova Vet Ketone/Glucose Meter and has a normal measurement range. Quality Control testing is performed exactly the same as blood testing. The result is available in 10 seconds and is automatically recorded into the meter’s memory. 1 bottle.
Catalog No. NC2882378