Learn More
Apexbio Technology LLC Ketoprofen, 1g Cas# 22071-15-4
SDP
Supplier:  Apexbio Technology LLC B14461000
Encompass_Preferred
Ketoprofen (Actron) is a non-selective NSAID with IC50 of 0.5 uM and 2.33 uM for human recombinant COX-1 and COX-2, respectively. 22071-15-4. MFCD00055790 Ketoprofen (Actron) is a non-selective NSAID with IC50 of 0.5 M and 2.33 M for human recombinant COX-1 and COX-2, respectively. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8617