Learn More
Apexbio Technology LLC Ketoprofen 22071-15-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062284
Encompass_Preferred
Ketoprofen (CAS 22071-15-4) is a non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase enzymes (COX) reducing prostaglandin synthesis and subsequent inflammation In biochemical assays using human recombinant enzymes ketoprofen demonstrates non-selective COX inhibition with reported IC50 values of approximately 0 5 M for COX-1 and 2 33 M for COX-2 Widely utilized in biomedical research ketoprofen serves as a pharmacological tool to investigate inflammatory signaling pathways and evaluate COX-dependent processes
Catalog No. 50-000-62284 Supplier Apexbio Technology LLC Supplier No. B14465.1
 Restricted Item
Add to Cart
Add to Cart
Ketoprofen (CAS 22071-15-4) is a non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase enzymes (COX) reducing prostaglandin synthesis and subsequent inflammation In biochemical assays using human recombinant enzymes ketoprofen demonstrates non-selective COX inhibition with reported IC50 values of approximately 0 5 M for COX-1 and 2 33 M for COX-2 Widely utilized in biomedical research ketoprofen serves as a pharmacological tool to investigate inflammatory signaling pathways and evaluate COX-dependent processes