Learn More
Selleck Chemical LLC Ketoprofen-50mg
SDP
Catalog No. 502389798
Encompass_Preferred
Ketoprofen (RP-19583) is a dual COX1/2 inhibitor, used as a nonsteroidal anti-inflammatory drug to treat arthritis-related inflammatory pains.
Catalog No. 50-238-9798 Supplier Selleck Chemical LLC Supplier No. S164550MG
Ketoprofen (RP-19583) is a dual COX1/2 inhibitor, used as a nonsteroidal anti-inflammatory drug to treat arthritis-related inflammatory pains.