Learn More
Medchemexpress LLC Ketoprofen-d3 | 159490-55-8 | 99.4% | 5 MG
SDP
Catalog No. 5000405498
Encompass_Preferred
Ketoprofen-d3 is the deuterium labeled Ketoprofen. Ketoprofen (RP-19583) is a non-steroidal antiinflammatory agent, acting as a potent inhibitor of COX, with IC50s of 2 nM and 26 nM for COX-1 and COX-2 in human blood monocytes, respectively. It is for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis.
  • Deuterium labeled compound
  • Non-steroidal anti-inflammatory agent
  • Potent inhibitor of COX-1 and COX-2
  • Used as a tracer for quantitation during drug development
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05498 Supplier Medchemexpress LLC Supplier No. HYB0227S5MG
Ketoprofen-d3 is the deuterium labeled Ketoprofen. Ketoprofen (RP-19583) is a non-steroidal antiinflammatory agent, acting as a potent inhibitor of COX, with IC50s of 2 nM and 26 nM for COX-1 and COX-2 in human blood monocytes, respectively. It is for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis.
  • Deuterium labeled compound
  • Non-steroidal anti-inflammatory agent
  • Potent inhibitor of COX-1 and COX-2
  • Used as a tracer for quantitation during drug development
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS