Learn More
Medchemexpress LLC Ketoprofen-d3 | 159490-55-8 | 99.8% | 1 MG
SDP
Catalog No. 5000406371
Encompass_Preferred
Ketoprofen-d3 is a deuterium labeled non-steroidal anti-inflammatory agent and a potent inhibitor of COX, with IC50s of 2 nM for COX-1 and 26 nM for COX-2 in human blood monocytes. It serves as a valuable tool for research involving quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06371 Supplier Medchemexpress LLC Supplier No. HYB0227S1MG
Ketoprofen-d3 is a deuterium labeled non-steroidal anti-inflammatory agent and a potent inhibitor of COX, with IC50s of 2 nM for COX-1 and 26 nM for COX-2 in human blood monocytes. It serves as a valuable tool for research involving quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS