Learn More
Medchemexpress LLC Ketoprofen-d4 | 1219805-29-4 | 98.7% | 1 MG
SDP
Catalog No. 5000405698
Encompass_Preferred
Ketoprofen-d4 (RP-19583-d4) is a deuterium-labeled derivative of Ketoprofen, a non-steroidal anti-inflammatory agent (NSAID). This compound acts as a potent inhibitor of cyclooxygenase (COX) enzymes, with reported IC50 values of 2 nM for COX-1 and 26 nM for COX-2 in human blood monocytes. Deuteration can influence the pharmacokinetic and metabolic profiles of drugs. Ketoprofen-d4 is suitable for use as a tracer in research applications and as an internal standard for quantitative analysis.
  • Potent inhibitor of cyclooxygenase enzymes
  • Suitable for use as a tracer in research applications
  • Ideal as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotope
  • Highly soluble in DMSO
Catalog No. 50-004-05698 Supplier Medchemexpress LLC Supplier No. HYB0227S11MG
Ketoprofen-d4 (RP-19583-d4) is a deuterium-labeled derivative of Ketoprofen, a non-steroidal anti-inflammatory agent (NSAID). This compound acts as a potent inhibitor of cyclooxygenase (COX) enzymes, with reported IC50 values of 2 nM for COX-1 and 26 nM for COX-2 in human blood monocytes. Deuteration can influence the pharmacokinetic and metabolic profiles of drugs. Ketoprofen-d4 is suitable for use as a tracer in research applications and as an internal standard for quantitative analysis.
  • Potent inhibitor of cyclooxygenase enzymes
  • Suitable for use as a tracer in research applications
  • Ideal as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotope
  • Highly soluble in DMSO