Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketoprofen, Micronized, USP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18603646 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-646 500 g Poly Bottle
18-603-644 25 g Poly Bottle
18-603-645 100 g Poly Bottle
18-603-647 1 kg Poly Bottle
18-603-648 5 kg Poly Bottle
18-603-649 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-603-646 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. K1392500GMPL
Add to Cart
Add to Cart
Request a Quote

K1392, 22071-15-4, C16H14O3

Spectrum™ Chemical Ketoprofen, Micronized, USP

Chemical Identifiers

CAS 22071-15-4
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.29
InChI Key DKYWVDODHFEZIM-UHFFFAOYNA-N
IUPAC Name 2-(3-benzoylphenyl)propanoic acid
SMILES CC(C(O)=O)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1

Specifications

CAS Max % 100%
Loss on Drying 0.5%
Melting Range 92.0-97.0°C
Packaging Poly Bottle
Quantity 500 g
UN Number UN2811
Specific Rotation +1° to -1°
Percent Purity 98.5 to 101%
Grade USP
Physical Form Micronized Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.