Learn More
Cayman Chemical Ketorolac, 74103-06-3, 100 mg
SDP
Supplier:  Cayman Chemical 9001148100
Encompass_Preferred
An NSAID, non-selective COX inhibitor, being marketed as an analgesic to manage postoperative pain as well as an ophthalmic solution to treat ocular pain and inflammation; inhibits COX-1 and COX-2 (IC50s = 20 nM) in vivo
Catalog No. NC0290450