Learn More
TARGETMOL CHEMICALS INC Ketorolac 1G
SDP
Supplier:  TARGETMOL CHEMICALS INC T214121G
Encompass_Preferred
Also available in 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Ketorolac (Acuvail) is a non-steroidal anti-inflammatory drug (NSAID) in the family of heterocyclic acetic acid derivatives. It acts by inhibiting the bodily synthesis of prostaglandins. Purity 99.68%
Catalog No. 50-287-9130