Learn More
Medchemexpress LLC Ketorolac | 74103-06-3 | 50 MG
SDP
Catalog No. 5000407867
Encompass_Preferred
Ketorolac (Standard) is the analytical standard of Ketorolac. It is a non-steroidal anti-inflammatory drug (NSAID) that acts as a nonselective COX inhibitor. It has IC50s of 20 nM for COX-1 and 120 nM for COX-2. Ketorolac tromethamine is used in a 0.5% ophthalmic solution for research into allergic conjunctivitis, cystoid macular edema, intraoperative miosis, and postoperative ocular inflammation and pain. Additionally, Ketorolac tromethamine is a DDX3 inhibitor and has potential for cancer research.
  • Analytical standard for Ketorolac
  • Non-steroidal anti-inflammatory drug (NSAID)
  • Acts as a nonselective COX inhibitor
  • Potential for research into allergic conjunctivitis, cystoid macular edema, intraoperative miosis, and postoperative ocular inflammation and pain
  • Functions as a DDX3 inhibitor with potential for cancer research
  • Intended for research and analytical applications
Catalog No. 50-004-07867 Supplier Medchemexpress LLC Supplier No. HYB0580R50MG
Ketorolac (Standard) is the analytical standard of Ketorolac. It is a non-steroidal anti-inflammatory drug (NSAID) that acts as a nonselective COX inhibitor. It has IC50s of 20 nM for COX-1 and 120 nM for COX-2. Ketorolac tromethamine is used in a 0.5% ophthalmic solution for research into allergic conjunctivitis, cystoid macular edema, intraoperative miosis, and postoperative ocular inflammation and pain. Additionally, Ketorolac tromethamine is a DDX3 inhibitor and has potential for cancer research.
  • Analytical standard for Ketorolac
  • Non-steroidal anti-inflammatory drug (NSAID)
  • Acts as a nonselective COX inhibitor
  • Potential for research into allergic conjunctivitis, cystoid macular edema, intraoperative miosis, and postoperative ocular inflammation and pain
  • Functions as a DDX3 inhibitor with potential for cancer research
  • Intended for research and analytical applications