Learn More
Medchemexpress LLC Ketorolac-d5 | 1215767-66-0 | 98.8% | 5 MG
SDP
Catalog No. 5000408462
Encompass_Preferred
Ketorolac-d5 is a deuterium labeled Ketorolac. Ketorolac is a non-steroidal anti-inflammatory agent, acting as a nonselective COX inhibitor, with IC50s of 20 nM for COX-1 and 120 nM for COX-2.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Store powder at -20°C for 3 years; 4°C for 2 years.
  • Store in solvent and stock solution at -80°C for 6 months; -20°C for 1 month.
Catalog No. 50-004-08462 Supplier Medchemexpress LLC Supplier No. HYB0580S5MG
Ketorolac-d5 is a deuterium labeled Ketorolac. Ketorolac is a non-steroidal anti-inflammatory agent, acting as a nonselective COX inhibitor, with IC50s of 20 nM for COX-1 and 120 nM for COX-2.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Store powder at -20°C for 3 years; 4°C for 2 years.
  • Store in solvent and stock solution at -80°C for 6 months; -20°C for 1 month.