Learn More
Medchemexpress LLC HY-B0580S 1mg Medchemexpress, Ketorolac D5 CAS:1215767-66-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0580S1MG
Encompass_Preferred
Medchemexpress, HY-B0580S 1mg Ketorolac D5 CAS:1215767-66-0 Ketorolac D5 is a deuterium labeled Ketorolac. Ketorolac is a non-steroidal anti-inflammatory agent, acting as a nonselective COX inhibitor, with IC50s of 20 nM for COX-1 and 120 nM for COX-2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2609