Learn More
U.S. Pharmacopeia Ketorolac Related Compound A, 167105-80-8, MFCD27978189, 20 mg
Catalog No. NC1363696
Encompass
Empirical Formula (Hill Notation): C19H22N2O5, Molecular Weight: 358.39, Synonym: 5-Benzoyl-N-(1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)-2,3-dihydro-1H-pyrrolizine-1-carboxamide.
Catalog No. NC1363696 Supplier U.S. Pharmacopeia Supplier No. 1356315
Empirical Formula (Hill Notation): C19H22N2O5, Molecular Weight: 358.39, Synonym: 5-Benzoyl-N-(1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)-2,3-dihydro-1H-pyrrolizine-1-carboxamide.