Learn More
U.S. Pharmacopeia Ketorolac Related Compound B, 154476-25-2, MFCD28411605, 20 mg
Catalog No. NC1363694
Encompass
Empirical Formula (Hill Notation): C14H13NO2, Molecular Weight: 227.26, Synonym: 5-Benzoyl-2,3-dihydro-1H-pyrrolizin-1-ol.
Catalog No. NC1363694 Supplier U.S. Pharmacopeia Supplier No. 1356326
Empirical Formula (Hill Notation): C14H13NO2, Molecular Weight: 227.26, Synonym: 5-Benzoyl-2,3-dihydro-1H-pyrrolizin-1-ol.