Learn More
Medchemexpress LLC HY-B0138 500mg Medchemexpress, Ketorolac (tromethamine salt) CAS:74103-07-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0138500MG
Encompass_Preferred
Medchemexpress, HY-B0138 500mg Ketorolac (tromethamine salt) CAS:74103-07-4 Ketorolac tromethamine salt (RS37619 tromethamine salt) is a non-steroidal anti-inflammatory agent, acting as a nonselective COX inhibitor, with IC50s of 20 nM for COX-1 and 120 nM for COX-2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2379