Learn More
Medchemexpress LLC Ketorolac (tromethamine salt) | 74103-07-4 | 99.5% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB013810MM/1ML
Encompass_Preferred
Ketorolac tromethamine salt is a non-steroidal anti-inflammatory agent (NSAIA) that acts as a nonselective COX inhibitor. It is provided as a 10 mM solution in 1 mL of DMSO, suitable for laboratory research. It is stable under recommended storage conditions, with specific handling guidelines to ensure its integrity.
  • Nonselective COX inhibitor
  • IC50s of 20 nM for COX-1 and 120 nM for COX-2
  • Purity of 99.53%
  • Supplied as a 10 mM solution in DMSO
  • Stable under recommended storage conditions
  • Causes skin and eye irritation
  • May cause respiratory irritation
Catalog No. 50-004-50692