Learn More
Medchemexpress LLC Ketorolac (tromethamine salt) | 74103-07-4 | MFCD00887595 | 99.5% | 1 G
SDP
Catalog No. 5000450949
Encompass_Preferred
Ketorolac tromethamine salt is a non-steroidal anti-inflammatory agent that acts as a nonselective COX inhibitor. With IC50s of 20 nM for COX-1 and 120 nM for COX-2, this compound is intended for laboratory chemical use and substance manufacturing, and is for research use only.
  • Non-steroidal anti-inflammatory agent
  • Nonselective COX inhibitor
  • Solid appearance
  • Purity of 99.5%
  • IC50 of 20 nM for COX-1
  • IC50 of 120 nM for COX-2
Catalog No. 50-004-50949 Supplier Medchemexpress LLC Supplier No. HYB01381G
Ketorolac tromethamine salt is a non-steroidal anti-inflammatory agent that acts as a nonselective COX inhibitor. With IC50s of 20 nM for COX-1 and 120 nM for COX-2, this compound is intended for laboratory chemical use and substance manufacturing, and is for research use only.
  • Non-steroidal anti-inflammatory agent
  • Nonselective COX inhibitor
  • Solid appearance
  • Purity of 99.5%
  • IC50 of 20 nM for COX-1
  • IC50 of 120 nM for COX-2