Learn More
Cayman Chemical Ketotifen-13C-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2748522-40-7, 1 mg
SDP
Catalog No. 502940281
Encompass_Preferred
An internal standard for the quantification of ketotifen by GC- or LC-MS
Catalog No. 50-294-0281 Supplier Cayman Chemical Supplier No. 288021 MG
An internal standard for the quantification of ketotifen by GC- or LC-MS