Learn More
Medchemexpress LLC Ketotifen | 34580-13-7 | 99.7% | 10 MM * 1 ML
SDP
Catalog No. 5000406083
Encompass_Preferred
Ketotifen is an orally active second-generation noncompetitive histamine 1 (H1) receptor blocker and mast cell stabilizer. It can block 6-phosphogluconate dehydrogenase (PGD) in vitro and also has antiviral activity against SARS-CoV-2 and Influenza virus. Ketotifen can be used for research on autoimmune encephalomyelitis (EAE) and asthma attack prevention.
  • Orally active second-generation noncompetitive histamine 1 (H1) receptor blocker and mast cell stabilizer.
  • Can block 6-phosphogluconate dehydrogenase (PGD) in vitro.
  • Has antiviral activity against SARS-CoV-2 and influenza virus.
  • Can be used for research of autoimmune encephalomyelitis (EAE) and asthma attack prevention.
Catalog No. 50-004-06083 Supplier Medchemexpress LLC Supplier No. HYB015710MM/1ML
Ketotifen is an orally active second-generation noncompetitive histamine 1 (H1) receptor blocker and mast cell stabilizer. It can block 6-phosphogluconate dehydrogenase (PGD) in vitro and also has antiviral activity against SARS-CoV-2 and Influenza virus. Ketotifen can be used for research on autoimmune encephalomyelitis (EAE) and asthma attack prevention.
  • Orally active second-generation noncompetitive histamine 1 (H1) receptor blocker and mast cell stabilizer.
  • Can block 6-phosphogluconate dehydrogenase (PGD) in vitro.
  • Has antiviral activity against SARS-CoV-2 and influenza virus.
  • Can be used for research of autoimmune encephalomyelitis (EAE) and asthma attack prevention.