Learn More
Medchemexpress LLC Ketotifen-d3 fumarate | 1795138-23-6 | 428.52 | C23H20D3NO5S | 10 MG
SDP
Catalog No. 5000298250
Encompass_Preferred
Ketotifen-d3 fumarate is the deuterium-labeled fumarate salt of ketotifen, provided as a research reagent and stable isotope for analytical applications. It functions as a noncompetitive H1-antihistamine and mast cell stabilizer and is intended for laboratory use only.
  • Deuterium-labeled internal standard for LC-MS quantification.
  • Fumarate salt form suitable for method development and tracer studies.
  • Available in small, precise pack sizes for analytical workflows.
  • Intended for research use only; not for clinical or diagnostic use.
Catalog No. 50-002-98250 Supplier Medchemexpress LLC Supplier No. HYB0157AS10MG
Ketotifen-d3 fumarate is the deuterium-labeled fumarate salt of ketotifen, provided as a research reagent and stable isotope for analytical applications. It functions as a noncompetitive H1-antihistamine and mast cell stabilizer and is intended for laboratory use only.
  • Deuterium-labeled internal standard for LC-MS quantification.
  • Fumarate salt form suitable for method development and tracer studies.
  • Available in small, precise pack sizes for analytical workflows.
  • Intended for research use only; not for clinical or diagnostic use.