Learn More
Medchemexpress LLC Ketotifen-d3 fumarate | 1795138-23-6 | 99.2% | 1 MG
SDP
Catalog No. 5000407163
Encompass_Preferred
Ketotifen-d3 (fumarate) is the deuterium labeled Ketotifen fumarate. Ketotifen (HC 20511) fumarate is a second-generation noncompetitive H1-antihistamine and mast cell stabilizer, which is used to prevent asthma attacks. It is for research use only and not sold to patients.
  • Deuterium labeled Ketotifen fumarate
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-07163 Supplier Medchemexpress LLC Supplier No. HYB0157AS1MG
Ketotifen-d3 (fumarate) is the deuterium labeled Ketotifen fumarate. Ketotifen (HC 20511) fumarate is a second-generation noncompetitive H1-antihistamine and mast cell stabilizer, which is used to prevent asthma attacks. It is for research use only and not sold to patients.
  • Deuterium labeled Ketotifen fumarate
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.