Learn More
Medchemexpress LLC Ketotifen-d3 fumarate | 1795138-23-6 | 99.2% | 5 MG
SDP
Catalog No. 5000406941
Encompass_Preferred
Ketotifen-d3 fumarate is a deuterium labeled version of Ketotifen fumarate, a second-generation noncompetitive H1-antihistamine and mast cell stabilizer. It is primarily used for research purposes, including as a tracer and internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS.
  • Deuterium labeled version for research applications
  • Functions as a noncompetitive H1-antihistamine
  • Acts as a mast cell stabilizer
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
Catalog No. 50-004-06941 Supplier Medchemexpress LLC Supplier No. HYB0157AS5MG
Ketotifen-d3 fumarate is a deuterium labeled version of Ketotifen fumarate, a second-generation noncompetitive H1-antihistamine and mast cell stabilizer. It is primarily used for research purposes, including as a tracer and internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS.
  • Deuterium labeled version for research applications
  • Functions as a noncompetitive H1-antihistamine
  • Acts as a mast cell stabilizer
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications