Learn More
Medchemexpress LLC Ketotifen-d3 (fumarate) | 1795138-23-6 | 99.23% | 25 MG
SDP
Catalog No. 5000407060
Encompass_Preferred
Ketotifen-d3 (fumarate) is the deuterium labeled form of Ketotifen fumarate. This compound acts as a second-generation noncompetitive H1-antihistamine and mast cell stabilizer. It is primarily used to prevent asthma attacks and is intended for research purposes only.
  • Deuterium labeled compound
  • Functions as a noncompetitive H1-antihistamine
  • Acts as a mast cell stabilizer
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis (e.g., NMR, GC-MS, LC-MS)
Catalog No. 50-004-07060 Supplier Medchemexpress LLC Supplier No. 25MGHYB0157AS
Ketotifen-d3 (fumarate) is the deuterium labeled form of Ketotifen fumarate. This compound acts as a second-generation noncompetitive H1-antihistamine and mast cell stabilizer. It is primarily used to prevent asthma attacks and is intended for research purposes only.
  • Deuterium labeled compound
  • Functions as a noncompetitive H1-antihistamine
  • Acts as a mast cell stabilizer
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis (e.g., NMR, GC-MS, LC-MS)