Learn More
Medchemexpress LLC Ketotifen (fumarate) | 34580-14-8 | 99.9% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB0157A1G
Encompass_Preferred
Ketotifen fumarate (HC 20-511) is an orally active, second-generation noncompetitive histamine 1 (H1) receptor blocker and mast cell stabilizer. It can block 6-phosphogluconate dehydrogenase (PGD) in vitro and exhibits antiviral activity against SARS-CoV-2 and Influenza virus. It is used in research for autoimmune encephalomyelitis (EAE) and asthma attack prevention.
  • Orally active, second-generation noncompetitive histamine 1 (H1) receptor blocker
  • Mast cell stabilizer
  • Blocks 6-phosphogluconate dehydrogenase (PGD) in vitro
  • Exhibits antiviral activity against SARS-CoV-2 and influenza virus
  • Used in research for autoimmune encephalomyelitis (EAE)
  • Used in research for asthma attack prevention
Catalog No. 50-004-07232