Learn More
Cayman Chemical Ketotifen (fumarate), Purity: ≥98%, CAS: 34580-14-8, 100 mg
SDP
Catalog No. 502984503
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 1.3 nM) and mast cell stabilizer; selective for H1 receptors over H2 and H3 receptors (Kis = 987 and 2,500 nM, respectively); inhibits compound 48/80-induced degranulation of rat peritoneal mast cells at 50 and 100 μM; inhibits the PCA reaction in rats by 54.6% at 20 mg/kg; inhibits the quick phase airway response in a rat ovalbumin-induced immediate airway response model at 30 mg/kg
Catalog No. 50-298-4503 Supplier Cayman Chemical Supplier No. 20303100 MG
A histamine H1 receptor antagonist (Ki = 1.3 nM) and mast cell stabilizer; selective for H1 receptors over H2 and H3 receptors (Kis = 987 and 2,500 nM, respectively); inhibits compound 48/80-induced degranulation of rat peritoneal mast cells at 50 and 100 μM; inhibits the PCA reaction in rats by 54.6% at 20 mg/kg; inhibits the quick phase airway response in a rat ovalbumin-induced immediate airway response model at 30 mg/kg