Learn More
Cayman Chemical Ketorolac (calcium salt), Purity: ≥98%, CAS: 167105-81-9, 500 mg
SDP
Catalog No. 502894534
Encompass_Preferred
An NSAID and COX inhibitor (IC50 = 20 nM for both COX-1 and COX-2); prevents increases in paw swelling, increases paw withdrawal latency in a hot-plate test, and decreases PGE2 levels in paw tissue in a mouse model of carrageenan-induced inflammation at 30 mg/kg; a racemic mixture of the active (S)-ketorolac and inactive (R)-ketorolac enantiomers
Catalog No. 50-289-4534 Supplier Cayman Chemical Supplier No. 9003475500 MG
An NSAID and COX inhibitor (IC50 = 20 nM for both COX-1 and COX-2); prevents increases in paw swelling, increases paw withdrawal latency in a hot-plate test, and decreases PGE2 levels in paw tissue in a mouse model of carrageenan-induced inflammation at 30 mg/kg; a racemic mixture of the active (S)-ketorolac and inactive (R)-ketorolac enantiomers