Learn More
Cayman Chemical KetrolactromethamIn salt 25g
SDP
Catalog No. 502895088
Encompass_Preferred
An NSAID and COX inhibitor (IC50s = 20 nM for COX-1 and COX-2); prevents increases in paw swelling, increases paw withdrawal latency in a hot-plate test, and decreases PGE2 levels in paw tissue in a mouse model of carrageenan-induced inflammation at 30 mg/kg; a racemic mixture of the active (S)-ketorolac and inactive (R)-ketorolac enantiomers
Catalog No. 50-289-5088 Supplier Cayman Chemical Supplier No. 7069025 G
An NSAID and COX inhibitor (IC50s = 20 nM for COX-1 and COX-2); prevents increases in paw swelling, increases paw withdrawal latency in a hot-plate test, and decreases PGE2 levels in paw tissue in a mouse model of carrageenan-induced inflammation at 30 mg/kg; a racemic mixture of the active (S)-ketorolac and inactive (R)-ketorolac enantiomers