Learn More
Medchemexpress LLC Kevetrin hydrochloride | 66592-89-0 | 98.0% | 179.67 | 1 ML
SDP
Catalog No. 5000392743
Encompass_Preferred
Kevetrin hydrochloride is a potent activator of p53, inducing apoptosis in TP53 wild-type and mutant acute myeloid leukemia cells. It exhibits preferential cytotoxic activity against blast cells.
  • Potent activator of p53.
  • Induces apoptosis in TP53 wild-type and mutant acute myeloid leukemia cells.
  • Exhibits preferential cytotoxic activity against blast cells.
Catalog No. 50-003-92743 Supplier Medchemexpress LLC Supplier No. HY1627110MM/1ML
Kevetrin hydrochloride is a potent activator of p53, inducing apoptosis in TP53 wild-type and mutant acute myeloid leukemia cells. It exhibits preferential cytotoxic activity against blast cells.
  • Potent activator of p53.
  • Induces apoptosis in TP53 wild-type and mutant acute myeloid leukemia cells.
  • Exhibits preferential cytotoxic activity against blast cells.