Learn More
Medchemexpress LLC Kevetrin hydrochloride | 66592-89-0 | 98.0% | 179.67 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY16271500MG
Encompass_Preferred
Kevetrin hydrochloride is a potent activator of p53. It induces apoptosis in both TP53 wild-type and mutant acute myeloid leukemia cells, exhibiting preferential cytotoxic activity against blast cells.
  • Potent activator of p53.
  • Induces apoptosis in TP53 wild-type and mutant acute myeloid leukemia cells.
  • Shows preferential cytotoxic activity against blast cells.
Catalog No. 50-003-92233