Learn More
Medchemexpress LLC Kevetrin hydrochloride | 66592-89-0 | 98.0% | 179.67 | 50MG
SDP
Catalog No. 5000392125
Encompass_Preferred
Kevetrin hydrochloride is a potent activator of p53, inducing apoptosis in both TP53 wild-type and mutant acute myeloid leukemia cells. This compound demonstrates preferential cytotoxic activity against blast cells and is intended for research use only.
  • Potent activator of p53.
  • Induces apoptosis in TP53 wild-type and mutant acute myeloid leukemia cells.
  • Exhibits preferential cytotoxic activity against blast cells.
  • Significantly inhibits KASUMI-1 cell growth.
  • Induces metallothionein (MT) expression in acute myeloid leukemia cells.
  • Increases p53 mRNA and protein levels, and induces p21 protein production.
  • Inhibits tumor growth and extends survival time in xenograft models.
Catalog No. 50-003-92125 Supplier Medchemexpress LLC Supplier No. HY1627150MG
Kevetrin hydrochloride is a potent activator of p53, inducing apoptosis in both TP53 wild-type and mutant acute myeloid leukemia cells. This compound demonstrates preferential cytotoxic activity against blast cells and is intended for research use only.
  • Potent activator of p53.
  • Induces apoptosis in TP53 wild-type and mutant acute myeloid leukemia cells.
  • Exhibits preferential cytotoxic activity against blast cells.
  • Significantly inhibits KASUMI-1 cell growth.
  • Induces metallothionein (MT) expression in acute myeloid leukemia cells.
  • Increases p53 mRNA and protein levels, and induces p21 protein production.
  • Inhibits tumor growth and extends survival time in xenograft models.