Learn More
Cayman Chemical Kevetrin (hydrochloride), Purity: ≥95%, CAS: 66592-89-0, 10 mg
SDP
Catalog No. 502937094
Encompass_Preferred
An activator of p53; increases levels of activated p53 and expression of p21 in A549 cells and decreases expression of the transcription factor E2F1 in a variety of cell lines; decreases cell viability in a panel of cancer cell lines (mean IC50 = 0.49 μM); reduces tumor growth in various mouse xenograft models at 200 mg/kg
Catalog No. 50-293-7094 Supplier Cayman Chemical Supplier No. 2828810 MG
An activator of p53; increases levels of activated p53 and expression of p21 in A549 cells and decreases expression of the transcription factor E2F1 in a variety of cell lines; decreases cell viability in a panel of cancer cell lines (mean IC50 = 0.49 μM); reduces tumor growth in various mouse xenograft models at 200 mg/kg