Learn More
Braintree Scientific Aramid Glove Small
SDP
Catalog No. 501954990
Encompass
Aramid Goatskin Handling Gloves. Black deerskin leather glove, lined with 100% fine knit Aramid. May be worn on their own for handling small animals. Fabricated for flexibility, comfort and safety during long periods of wear. Sold as pairs. Please note these gloves run large. **Sold for research purposes only and not for human use**
This item is not returnable. View return policy
Catalog No. 50-195-4990 Supplier Braintree Scientific Supplier No. KVLLTHS
Aramid Goatskin Handling Gloves. Black deerskin leather glove, lined with 100% fine knit Aramid. May be worn on their own for handling small animals. Fabricated for flexibility, comfort and safety during long periods of wear. Sold as pairs. Please note these gloves run large. **Sold for research purposes only and not for human use**