Learn More
MSC PRO-SAFE Sleeve, Kevlar, ANSI Cut A3, 14.0 in, 1 ea
Catalog No. 502886275
Encompass
This general purpose protective sleeve is crafted from Kevlar, designed to offer superior protection in environments requiring cut and abrasion resistance.
  • Made from Kevlar material
  • Features a thumb hole
  • Elastic openings at both ends for secure fit
  • Cut-resistant
  • ANSI/ISEA abrasion resistance level 1
  • ANSI/ISEA cut resistance level A3
  • Overall length 14.0 in
Catalog No. 50-288-6275 Supplier MSC Supplier No. 79139341
This general purpose protective sleeve is crafted from Kevlar, designed to offer superior protection in environments requiring cut and abrasion resistance.
  • Made from Kevlar material
  • Features a thumb hole
  • Elastic openings at both ends for secure fit
  • Cut-resistant
  • ANSI/ISEA abrasion resistance level 1
  • ANSI/ISEA cut resistance level A3
  • Overall length 14.0 in