Learn More
Sklar Instruments Kevorkian Biopsy Punch, Sklar, 11-1/2 in., 3mm x 8mm Bite, Rectangular Bite with 4 Lower Teeth, Straight, Stainless Steel. EA/1.
SDP
Catalog No. 501436835 Shop All Sklar Instruments Products
Encompass_Preferred
Kevorkian Biopsy Punch, Sklar, 11-1/2 in., 3mm x 8mm Bite, Rectangular Bite with 4 Lower Teeth, Straight, Stainless Steel. Sklar's Kevorkian Biopsy Punch is commonly used during colposcopy procedures to excise tissue from the endocervix for biopsy. This product has a 3mm x 8mm rectangular bite with 4 lower teeth. There is a basket to catch the tissue sample after excision. The non-ratcheted, spring style handle features one large ring that can fit multiple fingers and has a horn on the upper handle for thumb placement. The length of the shaft is 11-1/2 inches.
This item is not returnable. View return policy
Catalog No. 50-143-6835 Supplier Sklar Instruments Supplier No. 907540
Kevorkian Biopsy Punch, Sklar, 11-1/2 in., 3mm x 8mm Bite, Rectangular Bite with 4 Lower Teeth, Straight, Stainless Steel. Sklar's Kevorkian Biopsy Punch is commonly used during colposcopy procedures to excise tissue from the endocervix for biopsy. This product has a 3mm x 8mm rectangular bite with 4 lower teeth. There is a basket to catch the tissue sample after excision. The non-ratcheted, spring style handle features one large ring that can fit multiple fingers and has a horn on the upper handle for thumb placement. The length of the shaft is 11-1/2 inches.