Learn More
Euthanex Key Fill Bottle Adaptor - Isoflurane Only
Catalog No. 501540181
Encompass_Preferred
Key Fill Bottle Adaptor. Used to fill Vaporizer Well with Isoflurane. Direct connection with well assures that Isoflurane and vapors do not spill or get inhaled by user.
Catalog No. 50-154-0181 Supplier Euthanex Supplier No. EZM21
Key Fill Bottle Adaptor. Used to fill Vaporizer Well with Isoflurane. Direct connection with well assures that Isoflurane and vapors do not spill or get inhaled by user.