Learn More
PRODUCTION BASICS KEYBOARD DRAWER (WITH MOUSE TRAY)
Catalog No. NC1235813
Encompass_Preferred
Keyboard Drawer mounts to underside of any Production Basics workstation or your existing table. Slide out tray holds mouse on left or right side of drawer. Accommodates standard size keyboards and provides space for an ergonomic wrist rest.
Catalog No. NC1235813 Supplier PRODUCTION BASICS Supplier No. 8681
Keyboard Drawer mounts to underside of any Production Basics workstation or your existing table. Slide out tray holds mouse on left or right side of drawer. Accommodates standard size keyboards and provides space for an ergonomic wrist rest.