Learn More
Grainger Keyed Padlock 15/16 in Rectangle Blue
Catalog No. 502750091
Encompass_Preferred
The ABUS Padlock 63/64 in Vertical is a high security padlock made of hardened steel with a 14mm shackle. It features a precision pin cylinder with mushroom and spool pins for picking resistance. The body is 64mm wide and the shackle has a vertical clearance of 63mm. This padlock has an ABUS Plus disc cylinder which provides protection against lock manipulation. It can be used for securing gates, containers, machinery and more. The padlock body is zinc alloy with a black finish.
Catalog No. 50-275-0091 Supplier Grainger Supplier No. 14J871
Add to Cart
Add to Cart
The ABUS Padlock 63/64 in Vertical is a high security padlock made of hardened steel with a 14mm shackle. It features a precision pin cylinder with mushroom and spool pins for picking resistance. The body is 64mm wide and the shackle has a vertical clearance of 63mm. This padlock has an ABUS Plus disc cylinder which provides protection against lock manipulation. It can be used for securing gates, containers, machinery and more. The padlock body is zinc alloy with a black finish.