Learn More
Grainger Keyed Padlock 53/64 in Rectangle Gold
Catalog No. 502753079
Encompass_Preferred
The ABUS 55/64 in. Vertical Padlock is a high security padlock made of hardened steel with a 14mm shackle that provides moderate physical attack resistance. It features a precision pin tumbler cylinder with anti-drill protection and an ABUS Plus disc cylinder which provides over 130,000 key differs. This weatherproof padlock has a chrome-plated body and is rated for outdoor use. It can be keyed alike for convenience. The overall dimensions are 2.36 in. tall x 1.77 in. wide x 0.98 in. deep and it weighs 0.55 lbs.
Catalog No. 50-275-3079 Supplier Grainger Supplier No. 14J890
Add to Cart
Add to Cart
The ABUS 55/64 in. Vertical Padlock is a high security padlock made of hardened steel with a 14mm shackle that provides moderate physical attack resistance. It features a precision pin tumbler cylinder with anti-drill protection and an ABUS Plus disc cylinder which provides over 130,000 key differs. This weatherproof padlock has a chrome-plated body and is rated for outdoor use. It can be keyed alike for convenience. The overall dimensions are 2.36 in. tall x 1.77 in. wide x 0.98 in. deep and it weighs 0.55 lbs.