Learn More
Chroma Technologies KEYENCE BZX MICROSCOPE ET- CY7
SDP
Catalog No. NC1789777
Encompass_Preferred
Chroma filter sets mounted in cubes for Keyence BZX Microscope ET- CY7 ET710/75x ET810/90m T760lpxr-UF1
Catalog No. NC1789777 Supplier Chroma Technologies Supplier No. 49007UF1_91056
Chroma filter sets mounted in cubes for Keyence BZX Microscope ET- CY7 ET710/75x ET810/90m T760lpxr-UF1