Learn More
Keyence Corp Of America KEYENCE MK10 INK CARTRIDGE 2PK
Catalog No. NC2225987
Encompass
  • Two-pack of ink cartridges for high productivity.
  • Standard MK-10 ink formulation offers reliable marking.
  • Quick-drying ink with an approximate drying time of 1 sec.
  • Compatible with MK-G series continuous inkjet printers.
  • Suitable for a wide range of materials including metals, glass, and plastics.
  • Equipped with an IC chip for efficient expiration date management.
  • Not subject to PRTR regulations for safe industrial use.
  • 12 months shelf life after manufacture.
  • 12 months useful life after cartridge insertion.
This item is not returnable. View return policy
Catalog No. NC2225987 Supplier Keyence Corp Of America Supplier No. MKK02
  • Two-pack of ink cartridges for high productivity.
  • Standard MK-10 ink formulation offers reliable marking.
  • Quick-drying ink with an approximate drying time of 1 sec.
  • Compatible with MK-G series continuous inkjet printers.
  • Suitable for a wide range of materials including metals, glass, and plastics.
  • Equipped with an IC chip for efficient expiration date management.
  • Not subject to PRTR regulations for safe industrial use.
  • 12 months shelf life after manufacture.
  • 12 months useful life after cartridge insertion.