Learn More
Sklar Instruments Keyes Cutaneous Punch, Sklar 4 in., 4mm, Sharp, Straight, Fenestrated, Knurled, Stainless Steel. EA.
SDP
Catalog No. 501183982 Shop All Sklar Instruments Products
Encompass_Preferred
Keyes Cutaneous Punch, Sklar. 4 in., 4mm, Sharp, Straight, Fenestrated, Knurled, Stainless Steel. Merit Keyes Cutaneous Punch are used to collect a sample of tissue for biopsy. The punch is available in 2mm-6mm tips. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures.
Catalog No. 50-118-3982 Supplier Sklar Instruments Supplier No. 98404
Keyes Cutaneous Punch, Sklar. 4 in., 4mm, Sharp, Straight, Fenestrated, Knurled, Stainless Steel. Merit Keyes Cutaneous Punch are used to collect a sample of tissue for biopsy. The punch is available in 2mm-6mm tips. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures.