Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety KeyGuard™ Disposable Coverall
SDP

Catalog No. 19055577
This item is not returnable. View return policy
Catalog No. 19-055-577 Supplier Keystone Safety Supplier No. CVLKGE5XL

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Keyguard™ Microporous Coveralls deliver an ideal balance of protection, durability and comfort.

  • Keyguard™ (100% Polyolefin Tyvek™ Alternative)
  • Zipper Closure
  • Set-in Sleeves
  • Elasticized Cuffs
  • 100% latex free
  • Provides the work with outstanding protection, durability and comfort.

Specifications

Product Type Single-use Coveralls
Size 5X-Large
Gender Unisex
Material Microporous Fabric
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Snap Front
Seam Type Serged
Sterility Non-sterile
Latex Free Yes
No. of Pockets None
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.