Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Keyguard™ Disposable Pants
SDP

Catalog No. 19055300
Change view
Click to view available options
Size:
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-055-300 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-055-300 Supplier Keystone Safety Supplier No. PANTKGXL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pants protect workers from non-hazardous dry particulates. Worn over personal clothing to protect the bottom half of a worker from contaminants and dirt.

  • Disposable Pants
  • Microporous Fabric
  • 100% Latex Free
  • White
  • Seam Style: Serged
  • Waist Closure: Elastic

Specifications

Size X-Large
Gender Unisex
Material Keyguard
Color White
Ankle Style Open
Seam Type Serged
No. of Pockets None
Disposable Single-use
Latex Free Yes
Sterility Non-sterile
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.