Learn More
Keystone Safety Keyguard™ ISO Class 6 Microporous Gowns
SDP
Catalog No. 19990123
Change view
Click to view available options
2XL
Catalog No.
19-990-123 2XL
1 options
This item is not returnable. View return policy
Catalog No. 19-990-123 Supplier Keystone Safety Supplier No. ISOKGV2XL
This item is not returnable. View return policy
Manufactured from a specially formulated microporous material that makes these garments highly breathable and provides great comfort and protection
  • Low shedding, low in particulate, and antistatic to protect sensitive products
  • Excellent particulate barrier, liquid repellant, and wicks body moisture away to provide better comfort
Specifications
Size 2X-Large
Product Type Controlled Environments Gown
Color White
Disposable Single-use
Cuff Style Knit
Certifications/Compliance ISO Class 6