Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety KeyGuard™ Knit Wrist Sleeve
SDP

Catalog No. 19812909
Change view
Click to view available options
Construction:
Keyguard
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Construction
19-812-909 Keyguard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-812-909 Supplier Keystone Safety Supplier No. AGKG22
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cleanroom compatible, non-linting sleeve with sewn knit wrists is durable and comfortable.

  • Microporous
  • 100% Latex Free
  • Non-linting

Specifications

Product Type Controlled Environment Sleeve
Construction Keyguard
Color White
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Seam Type Serged
Latex Free Yes
Model AG-KG-22
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.