Learn More
Medicus Health Keyless Welded Storage Cabinets, Progammable Electronic Lock, Putty
SDP
Catalog No. NC3523384
Encompass
• Electronic Storage Cabinet is equipped with a keypad lock that keeps a record of the day, date and time of when the cabinet is accessed
• Cabinet lock also has a key override function, which enables you to manually unlock the device in any situations that may require emergency access
• Keyless code locking system offers secure storage and a programmable password
• Lock can store up to 8 open door records
• Cabinet lock also has a key override function, which enables you to manually unlock the device in any situations that may require emergency access
• Perfect for storing and locking up medical supplies
• Reinforced doors swing on three leaf hinges and a three-point door locking mechanism keeps contents safe
• Electronic storage cabinet comes with (2) adjustable shelves so you can customize storage needs
• Dimensions: 36"W x 18" D x 42"H
Catalog No. NC3523384 Supplier Medicus Health Supplier No. 2881M1
• Electronic Storage Cabinet is equipped with a keypad lock that keeps a record of the day, date and time of when the cabinet is accessed
• Cabinet lock also has a key override function, which enables you to manually unlock the device in any situations that may require emergency access
• Keyless code locking system offers secure storage and a programmable password
• Lock can store up to 8 open door records
• Cabinet lock also has a key override function, which enables you to manually unlock the device in any situations that may require emergency access
• Perfect for storing and locking up medical supplies
• Reinforced doors swing on three leaf hinges and a three-point door locking mechanism keeps contents safe
• Electronic storage cabinet comes with (2) adjustable shelves so you can customize storage needs
• Dimensions: 36"W x 18" D x 42"H